Learn More
Cole-Parmer™ Stuart™ Receiving Flask For Stuart Rotary Evaporator
Product Code. 12477802
Change view
Click to view available options
Volume (Metric):
50 mL
100 mL
Unit Size:
Pack of 2
Product Code. Volume (Metric) unitSize
12477802 100 mL Pack of 2
12497802 50 mL Pack of 2
2 options
Product Code. 12477802
Brand: Cole-Parmer™ 1009667
Only null left
Add to basket
Add to basket
Can be used with all Stuart™ glassware sets
  • For use with Stuart™ Rotary Evaporator
  • S35 spherical socket
  • Round bottom
Specifications
Type Receiving Flask
For Use With (Equipment) Stuart Rotary Evaporators
Shape Spherical
Taper S35 spherical socket
Volume (Metric) 100 mL