Learn More
Lenz Laborglasinstrumente™ Receiving Flask
Product Code. 11876134
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
2000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11876134 500 mL Each
11896104 100 mL Each
11826134 2000 mL Each
11806114 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11876134 Supplier Lenz Laborglasinstrumente™ Supplier No. 03063558
  • ONLINE EXCLUSIVE PRICE
    619.00 NOK
    434.99 NOK / Pack of 1
    Save 184.01 NOK
    29% Off
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Receiving Flasks are manufactured from durable DURAN™ glass.
  • With spherical joint socket S 35
  • With inscription label
  • Available in a range of capacities
Specifications
Material Duran Glass
Type Receiving Flask
For Use With (Equipment) Rotary Evaporators
Volume (Metric) 500 mL
Diameter (Metric) 105 mm