Learn More
Heidolph™ Receiving Flask
Product Code. 10660113 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
10 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10660113 500 mL Each
10005791 250 mL Each
10249601 1000 mL Each
10030972 10 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10660113 Supplier Heidolph™ Supplier No. 5148300000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 29-12-2026
For use with rotary evaporators
  • With S 35/20
  • Part number 15-300-004-11: includes drain valves
    • Included as a standard for all Hei-VAP Industrial models
Order Info
CapacityMetric
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Glass
Type Receiving Flask
For Use With (Equipment) Rotary evaporators
Standard Taper Joint Number Center 35/20
Volume (Metric) 500 mL