Learn More
Macherey-Nagel™ Receiver Plate
Product Code. 11942542
Click to view available options
Pore Size:
50 μm
Includes:
Filter Frits
Hydrophilized Filter Frits
Unit Size:
Pack of 4
Product Code. 11942542
Brand: Macherey-Nagel™ 740688.4
Use for filtration in 96-well format. Macherey Nagel™ Receiver Plates are important accessories for high throughput work.
  • Receiver plate accessory
  • High-throughput applications
  • Filter frits
  • Suitable for centrifugation and vacuum
Specifications
Pore Size 50 μm
Includes Filter Frits