Learn More
Reboxetine Mesylate, TRC
Product Code. 18072410
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072410 10 mg 10mg
30137619 25 mg 25mg
18022020 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18072410 Supplier Toronto Research Chemicals Supplier No. TRCR14200010MG
 Restricted Item
Estimated Shipment: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98769-84-7
Molecular Formula C19 H23 N O3 . C H4 O3 S
Molecular Weight (g/mol) 409.5
Synonym Reboxetine Mesilate, Morpholine, 2-[(2-ethoxyphenoxy)phenylmethyl]-, (R*,R*)-(±)-, methanesulfonate, Morpholine, 2-[(R)-(2-ethoxyphenoxy)phenylmethyl]-, (2R)-rel-, methanesulfonate (9CI), (±) Reboxetine mesylate, Davedax, Edronax, FCE 20124, PNU 155905E, PNU 155950E, Reboxetine mesilate, Reboxetine mesylate, Morpholine, 2-[(R)-(2-ethoxyphenoxy)phenylmethyl]-, (2R)-rel-, methanesulfonate (1:1)
IUPAC Name (2R)-2-[(R)-(2-ethoxyphenoxy)-phenylmethyl]morpholine;methanesulfonic acid
SMILES CCOc1ccccc1O[C@@H]([C@H]2CNCCO2)c3ccccc3.CS(=O)(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C19H23NO3.CH4O3S/c1-2-21-16-10-6-7-11-17(16)23-19(15-8-4-3-5-9-15)18-14-20-12-13-22-18;1-5(2,3)4/h3-11,18-20H,2,12-14H2,1H3;1H3,(H,2,3,4)/t18-,19-;/m1./s1
Formula Weight 409.1559
Chemical Name or Material Reboxetine Mesilate