missing translation for 'onlineSavingsMsg'
Learn More

Reboxetine mesylate, MedChemExpress

Product Code. p-200097429
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16456098 10 mg 10mg
16448697 5 mg 5mg
16442237 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 16456098

Brand: Medchem Express HY14560C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Reboxetine mesylate (FCE20124 mesylate) is a potent, selective, and specific noradrenaline reuptake inhibitor (NARI) for the research of depression. Reboxetine mesylate inhibits the uptake of norepinephrine, with a Ki of 8 nM.

Reboxetine mesylate (FCE20124 mesylate) is a potent, selective, and specific noradrenaline reuptake inhibitor (NARI) for the research of depression. Reboxetine mesylate inhibits the uptake of norepinephrine, with a Ki of 8 nM.

Specifications

Chemical Name or Material Reboxetine mesylate
CAS 98769-84-7
Color White
Purity Grade Notes Research
Molecular Formula C20H27NO6S
Quantity 10 mg
Synonym FCE20124 mesylate PNU155950E mesylate
Solubility Information DMSO : 125 mg/mL (305.25 mM; Need ultrasonic) ∣H2O : 50 mg/mL (122.10 mM; Need ultrasonic)
SMILES CS(=O)(O)=O.CCOC1=C(C=CC=C1)O[C@]([C@@]2([H])CNCCO2)([H])C3=CC=CC=C3.[relative stereochemistry]
Molecular Weight (g/mol) 409.5
Formula Weight 409.5
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.