Learn More
Heidolph™ Reax 20/40
Product Code. 10292351 Shop All Heidolph Instruments Products
Change view
Click to view available options
Speed Range:
1 to 16 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
10292351 1 to 16 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10292351 Supplier Heidolph™ Supplier No. 5412000400
  • ONLINE EXCLUSIVE PRICE
    42455.00 NOK
    41331.79 NOK / Pack of 1
    Save 1123.21 NOK
    2% Off
 Restricted Item
Estimated Shipment: 02-02-2027
Rotator Heidolph Reax 20/4 that features an analog control knob from 1 to 16 rpm that is ideal for waster water analysis.
  • Rotator Heidolph Reax 20/4 maximum tube diameter 136mm, maximum number of tubes 4, maximum speed 16rpm, with speed control
  • Set and continuously adjust the variable speed on the analog control knob from 1 to 16 rpm - ideal for waste water analysis
  • A model with a rotation speed of 2 to 32 rpm is available upon request
  • Easy and fast attachments of bottles
  • Other vessels with 160 - 270 mm height and max. 136 mm diameter can be used with attachments
  • All models have been designed with a low center of gravity which prevents gliding at high speed, even on a damp surface
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 1 to 16 rpm