Learn More
BD Biosciences Reagent Rack - PE
Product Code. 15991260
Change view
Click to view available options
Dimensions (L x W):
180 mm x 54.5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15991260 180 mm x 54.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15991260 Supplier BD Biosciences Supplier No. 657178
 Restricted Item
Estimated Shipment: 05-11-2026
Pre-cooled reagent rack keeps your antibodies cooled for 75 minutes, preserving their quality and protein-dye stability.
Pre-cooled BD™ Reagent Racks keep your antibodies cooled for 75 minutes, preserving their quality and protein-dye stability. The BD Reagent Racks are stackable and available in different colors to help you keep your vials organized on the bench as well as in the fridge. Each rack is filled with cooling material, and can accommodate up to 25 BD™ Amber vials and 20 narrower BD Pharmingen™ type vials. Size 180 mm x 54,5 mm - Weight: 950 gr.
TRUSTED_SUSTAINABILITY
Specifications
Holds BD Amber Vials (5) and Narrower BD Pharmingen Type Vials (20)
Stackable Yes
Dimensions (L x W) 180 mm x 54.5 mm
Weight (Metric) 950 G
Includes Filled with cooling material.