Specifications
| CAS | 17681-50-4 |
| Molecular Formula | C32H19ClN8Na4O14S4 |
| Synonym | C.I. 18105, Cibacron Brilliant Red 3B-A |
| InChI Key | HMBFFLCYQUKBIW-UHFFFAOYSA-J |
| SMILES | C1=CC=C(C=C1)C(=O)NC2=C3C(=CC(=C2)S(=O)(=O)[O-])C=C(C(=NNC4=C(C=CC(=C4)NC5=NC(=NC(=N5)NC6=CC=C(C=C6)S(=O)(=O)[O-])Cl)S(=O)(=O)[O-])C3=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+] |
| IUPAC Name | tetrasodium;5-benzamido-3-[[5-[[4-chloro-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate |
| Molecular Weight (g/mol) | 995.195 |
| PubChem CID | 44134921 |
| Formula Weight | 995.2 |
| Color | Purple to Red |
| Show More |