Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical
Product Code. 11542204
Click to view available options
Capacity (Metric):
0.25 L
1 L
6 L
10 L
20 L
Diameter (Metric) Outer:
85 mm
131 mm
236 mm
280 mm
350 mm
Includes:
Lab Flange 100 LF
Lab Flange 150 LF
Lab Flange 200 LF
Lab Flange 60 LF
Unit Size:
Each
Product Code. 11542204
Brand: Lenz Laborglasintrumente 06005594
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Without graduation
  • With laboratory flange LF
  • With groove
  • Made of DURAN™ tubing
  • Other sizes on request
Specifications
Capacity (Metric) 20 L
Material Duran Glass
Height (Metric) 385 mm
Includes Lab Flange 200 LF
No. per Pack 1 Ea.
Diameter (Metric) Outer 350 mm