Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical
Click to view available options
Capacity (Metric):
0.25 L
1 L
6 L
10 L
20 L
Diameter (Metric) Outer:
85 mm
131 mm
236 mm
280 mm
350 mm
Includes:
Lab Flange 100 LF
Lab Flange 150 LF
Lab Flange 200 LF
Lab Flange 60 LF
Unit Size:
Each
Description
- Without graduation
- With laboratory flange LF
- With groove
- Made of DURAN™ tubing
- Other sizes on request
Specifications
Specifications
| Capacity (Metric) | 6 L |
| Material | Duran Glass |
| Height (Metric) | 285 mm |
| Includes | Lab Flange 150 LF |
| No. per Pack | 1 Ea. |
| Diameter (Metric) Outer | 236 mm |