Learn More
Thermo Scientific™ Reacti-Vial™ Small Glass Reaction Vials
Product Code. 11742257 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.3 mL
1 mL
3 mL
5 mL
10 mL
Color:
Amber
Clear
Unit Size:
Pack of 12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Unit Size
11742257 0.3 mL Clear Pack of 12
11507350 1 mL Amber Pack of 12
11517350 5 mL Amber Pack of 12
11732257 0.1 mL Clear Pack of 12
11517360 1 mL Clear Pack of 12
11527360 3 mL Clear Pack of 12
11547360 10 mL Clear Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11742257 Supplier Thermo Scientific™ Supplier No. TS13220
 Restricted Item
Estimated Shipment: 08-10-2026
Use Thermo Scientific™ Reacti-Vial™ Small Reaction Vials in the Reacti-Therm™ Heating and Stirring module.
Use Thermo Scientific™ Reacti-Vial™ Small Reaction Vials in the Reacti-Therm™ Heating and Stirring module. An internal cone makes small sample handling in these vials easy and convenient. The extra thick glass wall magnifies the sample, making these vials ideal for observing chemical reactions. Reacti-Vials are available in a variety of sizes of clear and amber glass, and are supplied with Thermo Scientific™ Open-Top Screw Caps and PTFE/Rubber Discs.
Reacti-Vial™ Small Reaction Vials are ideal for:
  • Residue isolation
  • Derivative preparation
  • Maximum sample retrieval
  • Moisture protection
  • Sample storage
  • Precipitations
  • Centrifugations
  • Solvent evaporation
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Glass
Volume (Metric) 0.3 mL
Septum PTFE/Rubber
Thread Style Internal
Product Line Reacti-Vial
Volume (Metric) Total 0.3 mL
Closure Type Screw Cap (Open-top) with Septum
Quantity 12 Pk.
Closure Size 13-425
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Product Type Vial