Learn More
Thermo Scientific™ Gas Regulators for Reacti-Vap™ Evaporators
Product Code. 15224613 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
1/4 in. Tube Fitting
1/8 in. Tube Fitting
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
15224613 1/8 in. Tube Fitting Each
15234613 1/4 in. Tube Fitting Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15224613 Supplier Thermo Scientific™ Supplier No. 60181625
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Use these gas regulators to deliver the right amount of gas for sample evaporation using the Thermo Scientific™ ReactiVap™ Evaporator.
Deliver the right amount of gas to your Thermo Scientific™ ReactiVap™ Evaporator using these gas regulators. The Reacti-Vap Evaporator delivers pressurized gas to easily and efficiently evaporate samples for gas chromatography (GC) analysis. Choose between gas regulators with a 1/8 inch or a 1/4 inch tube fitting.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Reacti-Vap Evaporators
Includes 1/8 in. Tube Fitting
Product Type Gas Regulator