missing translation for 'onlineSavingsMsg'
Learn More

RDH13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16067-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16845544

  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Does not exhibit retinol dehydrogenase (RDH) activity in vitro.
Specifications

Specifications

RDH13
Polyclonal
Unconjugated
RDH13
RDH13, Retinol dehydrogenase 13, SDR7C3, UNQ736/PRO1430
Rabbit
Antigen Affinity Chromatography
RUO
108841, 112724, 361504
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CEE7, Q8NBN7
Rdh13
RDH13 Fusion Protein Ag8944
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers