Learn More
RDH10 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Product Code. 16803394 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16803394 20 μL 20µL
16893384 150 μL 150µL
2 options
Product Code. 16803394
Brand: Proteintech 146441AP20UL
Rabbit Polyclonal Antibody
The RDH10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RDH10. This antibody recognizes human, mouse, rat antigen. The RDH10 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Antigen RDH10
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration 300 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 157506
Target Species Human, Rat, Mouse
Isotype IgG