missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCAN1 Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-46853JF525
This item is not returnable.
View return policy
Description
RCAN1 Polyclonal antibody specifically detects RCAN1 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| RCAN1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 525 | |
| ADAPT78, calcipressin-1, calcium and oxidant-inducible mRNA, Down syndrome candidate region 1, Down syndrome critical region gene 1, Down syndrome critical region protein 1, DSC1, modulatory calcineurin-interacting protein 1, Myocyte-enriched calcineurin-interacting protein 1, near DSCR proline-rich protein, regulator of calcineurin 1CSP1DSCR1MCIP1RCN1 | |
| Synthetic peptide made to an internal portion of human RCAN1 (within residues 210-252). [Swiss-Prot# P53805] | |
| 0.1 mL | |
| Cancer, Neuroscience, Signal Transduction | |
| 1827 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction