Learn More
KNF™ RC600 and RC900 Rotary Evaporators PROMO
Description
- Shaped bath uses less fluid and energy
- Small footprint to save bench/fumehood space
- Easy fit, one handed flask attachment
- Cordless bath with spout and hand-holds for safety
- Simple intuitive controls for ease of use
- Memory function of flask height and rotation speed
- Optional 'intelligent' protective bath cover, can stop rotation and lift flask from bath if left open during operation
- Safe and easy adjustment of flask angle without danger of dropping condenser
- High efficiency condenser with anti chimney effect feature
- Quiet operation
- Automatic vacuum control if used with KNF SC 920 G Vacuum System
- Dry ice condenser available
Model Specific:
Model: RC600
- Fixed to tower for more usable bench space
Model: RC900
- Wireless remote control allows full operation from outside closed fume cupboard for safety and energy saving
- Tubing hidden inside for more usable bench space
- Adjustable alternating flask rotation for powder drying
Specifications
Specifications
| Description | Plug type: UK |
| Display | LCD |
| Includes | Without bath cover |
| Type | Rotary Evaporator |
| Model | RC600 |