Learn More
RBS™ Neutral Cleaning Agent
Product Code. 12883512
Change view
Click to view available options
Quantity:
20 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12883512 20 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12883512 Supplier RBS Supplier No. RBSVIRODRUM
Add to basket
Add to basket
Suitable for soaking
  • RB525 is foaming, with a pH of 10.3 when using 20g per L of water, suitable for soaking
  • RB535 is foaming, with a pH of 10.3 when using 20g per L of water, suitable for soaking and permits lengthy cleaning without calcium precipitation
  • RB550 is non-foaming, with bacterial action with a pH of 10.6 when using 10g per L of water, suitable for soaking and automatic washing machines
  • All three are suitable for ultrasonic cleaners
  • RBSVIRO is a neutral foaming, surfacing, non-toxic, non-irritant and non-corrosive agent with a pH of 7, suitable for soaking
Recommended for use in research, pathology, chemistry, biology, food and drink, pharmaceutical, electronics, optical and glass industry laboratories.
Specifications
Product Type Neutral Cleaning Agent
For Use With (Application) Suitable for Soaking
Quantity 20 kg
Volume (Metric) 20 kg