Learn More
RBS™ Alkaline Cleaning Agent
Click to view available options
Product Line:
RBS 25
RBS 35
RBS 50
Unit Size:
20 L
30kg
Each
Description
- RB525 is foaming, with a pH of 10.3 when using 20g per L of water, suitable for soaking
- RB535 is foaming, with a pH of 10.3 when using 20g per L of water, suitable for soaking and permits lengthy cleaning without calcium precipitation
- RB550 is non-foaming, with bacterial action with a pH of 10.6 when using 10g per L of water, suitable for soaking and automatic washing machines
- All three are suitable for ultrasonic cleaners
- RBSVIRO is a neutral foaming, surfacing, non-toxic, non-irritant and non-corrosive agent with a pH of 7, suitable for soaking
Specifications
Specifications
| Product Type | Alkaline Cleaning Agent |
| Product Line | RBS 50 |
| Appearance | Colorless |
| For Use With (Application) | Suitable for Soaking and Automatic Washing Machines |
| Quantity | 30 kg |