missing translation for 'onlineSavingsMsg'
Learn More

RBPMS Antibody, Novus Biologicals™

Product Code. 18789383 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. unitSize
18789383 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 18789383

Brand: Novus Biologicals NBP220112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 5 publications

RBPMS Polyclonal specifically detects RBPMS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.
TRUSTED_SUSTAINABILITY

Specifications

Applications Western Blot, Immunocytochemistry
Gene Alias FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing
Gene Symbols RBPMS
Immunogen Recombinant protein encompassing a sequence within the center region of human RBPMS. The exact sequence is proprietary.
Regulatory Status RUO

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.