missing translation for 'onlineSavingsMsg'
Learn More

RBMY1A1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14239-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892784

  • 1281.92 NOK / 20µL
Estimated Shipment: 20-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RBMY1A1 is a protein containing an RNA-binding motif in the N-terminus and four SRGY (serine, arginine, glycine, tyrosine) boxes in the C-terminus. This protein is thought to function as a splicing regulator during spermatogenesis. Multiple closely related paralogs of this gene are found in a gene cluster in the AZFb azoospermia factor region of chromosome Y. Most of these related copies are thought to be pseudogenes, though several likely encode functional proteins.
Specifications

Specifications

RBMY1A1
Polyclonal
Unconjugated
RBMY1A1
hRBMY, RBM1, RBM2, RBMY, RBMY1A1, RBMY1C, RNA binding motif protein 1, RNA binding motif protein 2, YRRM1, YRRM2
Rabbit
Antigen Affinity Chromatography
RUO
5940
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P0DJD3
RBMY1A1
RBMY1A1 Fusion Protein Ag5493
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers