missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10587-1-AP-20UL
This item is not returnable.
View return policy
Description
May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.Specifications
| RBM15 | |
| Polyclonal | |
| Unconjugated | |
| RBM15 | |
| One twenty two protein 1, OTT, OTT1, RBM15, RNA binding motif protein 15, SPEN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229700, 64783, 684233 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q0VBL3, Q96T37 | |
| RBM15 | |
| RBM15 Fusion Protein Ag0938 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction