missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RbAp46 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20365-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to RBBP7.
This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation.Specifications
RbAp46 | |
Polyclonal | |
Unconjugated | |
RBBP7 | |
Histone binding protein RBBP7, RbAp46, RBBP 7, RBBP7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
245688, 5931, 83712 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16576, Q60973, Q71UF4 | |
RBBP7 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |