missing translation for 'onlineSavingsMsg'
Learn More

Raxatrigine, MedChemExpress

Product Code. p-200101652
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16435358

Brand: Medchem Express HY12796

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Raxatrigine (GSK-1014802) is a novel small molecule state-dependent sodium channel blocker; Nav1.7 sodium channel inhibitor.

Raxatrigine (GSK-1014802) is a novel small molecule state-dependent sodium channel blocker

Specifications

Chemical Name or Material Raxatrigine
CAS 934240-30-9
Color White
Purity Grade Notes Research
Molecular Formula C18H19FN2O2
Quantity 10 mg
Synonym GSK-1014802 CNV1014802
Solubility Information DMSO : 83 mg/mL (264.04 mM; Need ultrasonic and warming)
SMILES O=C([C@H]1N[C@@H](C2=CC=C(OCC3=CC=CC=C3F)C=C2)CC1)N
Molecular Weight (g/mol) 314.35
Formula Weight 314.35
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.