Learn More
Raxatrigine, MedChemExpress
Product Code. 16435358
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16435358
Brand: Medchem Express HY12796
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Raxatrigine (GSK-1014802) is a novel small molecule state-dependent sodium channel blocker; Nav1.7 sodium channel inhibitor.
Raxatrigine (GSK-1014802) is a novel small molecule state-dependent sodium channel blocker
Specifications
Chemical Name or Material Raxatrigine
CAS 934240-30-9
Color White
Purity Grade Notes Research
Molecular Formula C18H19FN2O2
Quantity 10 mg
Synonym GSK-1014802 CNV1014802
Solubility Information DMSO : 83 mg/mL (264.04 mM; Need ultrasonic and warming)
SMILES O=C([C@H]1N[C@@H](C2=CC=C(OCC3=CC=CC=C3F)C=C2)CC1)N
Molecular Weight (g/mol) 314.35
Formula Weight 314.35
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.