missing translation for 'onlineSavingsMsg'
Learn More

Raxatrigine hydrochloride, MedChemExpress

Product Code. p-200101198
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16461547

Brand: Medchem Express HY12796A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Raxatrigine hydrochloride (GSK-1014802 hydrochloride) is a novel small molecule state-dependent sodium channel blocker; Nav1.7 sodium channel inhibitor.

Raxatrigine hydrochloride (GSK-1014802 hydrochloride) is a novel small molecule state-dependent sodium channel blocker

Specifications

Chemical Name or Material Raxatrigine hydrochloride
CAS 934240-31-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H20ClFN2O2
Quantity 50 mg
Synonym GSK-1014802 hydrochlorideCNV1014802 hydrochloride
Solubility Information DMSO : ≥ 31 mg/mL (88.36 mM) ∣H2O : 14.29 mg/mL (40.73 mM; Need ultrasonic)
SMILES O=C([C@H]1N[C@@H](C2=CC=C(OCC3=CC=CC=C3F)C=C2)CC1)N.[H]Cl
Molecular Weight (g/mol) 350.82
Formula Weight 350.82
Percent Purity 99.17%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.