Learn More
Reagecon™ Ratio Turbidity Standard
Product Code. 11429169 Shop All Reagecon Products
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
Turbidity:
0.2 NTU
2 NTU
20 NTU
50 NTU
90 NTU
Unit Size:
100mL
500mL
Product Code. Volume (Metric) Turbidity unitSize
11429169 500 mL 2 NTU 500mL
11536583 100 mL 50 NTU 100mL
11788455 100 mL 20 NTU 100mL
11449159 500 mL 0.2 NTU 500mL
11556583 100 mL 90 NTU 100mL
5 options
Product Code. 11429169
Brand: Reagecon™ CRSR2500
Specifications
Type Ratio Turbidity Standard
For Use With (Equipment) Ratio Instruments
Accuracy ±1%
Turbidity 2 NTU
Certifications/Compliance NIST, US EPA Approved
Volume (Metric) 500 mL