Learn More
Rat μ Opioid R/OPRM1 Antibody, R&D Systems™
Description
mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | mu Opioid R/OPRM1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1126D |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide |
| Gene Accession No. | P33535 |
| Gene Alias | hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1 |
| Gene Symbols | OPRM1 |
| Show More |
For Research Use Only