missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IL-13 Recombinant Protein

Product Code. 17616725
Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Unit Size:
100µg
10µg
25µg
500µg
This item is not returnable. View return policy

Product Code. 17616725

Brand: Invitrogen™ RP87927

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42203
Concentration 200 μg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation PBS with no preservative; pH 7.2
Gene ID (Entrez) 116553
Molecular Weight (g/mol) 12.3 kD
Name Rat IL-13 (aa 19-131)
Quantity 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration 0.01 ng/ μg cytokine
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; Interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Gene Symbol Il13
Specific Reactivity Rat
Conjugate Unconjugated
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.