missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat GM-CSF, AOF Recombinant Protein

Product Code. 17656585
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Product Code. Quantity unitSize
17656585 500 μg 500µg
17636585 100 μg 100µg
17646585 1 mg 1mg
3 options
This item is not returnable. View return policy

Product Code. 17656585

Brand: Invitrogen™ RP87782

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

GM-CSF (Granulocyte-Macrophage colony-stimulating factor) is a 14.6kDa hematopoietic growth factor that exists in glycosylated and non-glycosylated biologically active forms, and stimulates the development of granulocytes, macrophages, early megakaryocytes and eosinophil progenitor cells. The active form of the GM-CSF protein is found extracellularly as a homodimer and the GM-CSF gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q-syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. The ability of recombinant GM-CSF to increase hematopoietic cell recovery has become a focus area in the therapeutic treatment of patients following bone marrow transplantation. Recent studies have investigated GM-CSF in inflammatory and autoimmune diseases such as arthritis, arthritis related interstitial lung disease, nephritis, and psoriasis. In the CNS, GM-CSF depletion or neutralization has been studied in experimental autoimmune encephalomyelitis (EAE) and multiple sclerosis (MS).
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 128 aa.
  • Biological Activity: <20 pg/mL
  • Molecular Weight: 14.7 kDa
  • Animal Origin Free: Yes
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48750
For Use With (Application) Control, ELISA, Functional Assay, Gel Shift, Western Blot
Formulation 20 mM sodium bicarbonate with no preservative
Gene ID (Entrez) 116630
Molecular Weight (g/mol) 14.7 kDa
Name Rat GM-CSF, AOF
Quantity 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); Colony-stimulating factor; CSF; Csf2; Csfgm; GMCSF; GM-CSF; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; Granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; Molgramostin; put. GM-CSF; Sargramostim
Gene Symbol CSF2
Specific Reactivity Rat
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.