Learn More
RASEF Mouse anti-Human, Unconjugated, Clone: 1C2B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67159-1-IG-150UL
This item is not returnable.
View return policy
Description
The RASEF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RASEF. This antibody recognizes human antigen. The RASEF antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RASEF | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1C2B2 | |
| Unconjugated | |
| 150 μL | |
| 158158 | |
| IgG2a |