Learn More
RASEF Mouse anti-Human, Unconjugated, Clone: 1C2B2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67159-1-IG-20UL
 View more versions of this product
Product Code. 16849635
  • 1710.33 NOK / 20µL
Description
The RASEF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RASEF. This antibody recognizes human antigen. The RASEF antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RASEF
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1C2B2
Unconjugated
20 μL
158158
IgG2a