Learn More
Labconco™ RapidVap Vacuum Evaporation System with Lid Heater, 230 V
Product Code. 12624267 Shop All Labconco Corporation Products
Change view
Click to view available options
Volume (Metric):
0 to 3600 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12624267 0 to 3600 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12624267 Supplier Labconco™ Supplier No. 7900003
  • ONLINE EXCLUSIVE PRICE
    242867.82 NOK
    207396.15 NOK / Pack of 1
    Save 35471.67 NOK
    14% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-02-2027
Alternative to traditional centrifugal evaporation methods, evaporates using vortex motion, heat and vacuum
  • Processes up to 110 samples at once
  • Interchangeable aluminum sample blocks coated with PTFE
  • Hold tubes from 12 mm (outer diameter) to 600 mL
  • Microprocessor controls vacuum at preset level
    • Automatic vacuum release if power fails
  • LCD shows vacuum level from 0 to 999 mbar
  • Rear-mounted vacuum connections: vacuum port, tubing clamps, lid heater receptacle, tubing for connection to accessory vacuum pump
  • Lexan™ shield on clear glass lid protects against glass implosion
  • Dual lid clamps hold lid securely in place for vacuum-tight seal
  • Phenol-free gasket provides complete sealing under vacuum
  • PTFE-coated aluminum chamber
  • Audible/visual alarm for end point detected by temp sensors at heater and block
  • Optional 40 W lid heater plugs into electrical receptacle on rear
  • Eliminates fogging, increases visibility when evaporating water or other high-boiling-point solvents
  • Improved control panel
  • Easy-to-read LCD display prompts operator to program desired parameters and displays vortex speed by percentage, actual and set temperatures in °C, total time remaining in min.
    • Enter up to nine different protocols into memory for repetition of exact test parameters
  • Contamination and maintenance-free 1000 W dry block heating system speeds evaporation rate by supplying controlled amount of heat from ambient up to 100 °C
  • Heater and sample block move in tandem for more efficient heat transfer
  • Alarm signals completion of run, allowing samples to be left unattended while evaporating to desired endpoint
  • Brushless DC motor ensures safe, maintenance-free operation and provides smooth, variable-speed power up to 1000 rpm
  • Produces no potentially dangerous sparks
  • Systems may be used with broad range of strong chemicals
  • Powder-coated, aluminum cabinet base with smooth, solvent-resistant acrylic/PVC thermoplastic upper housing
  • Aluminum chamber with PTFE coating conducts heat efficiently and resists corrosion
  • All mechanical components isolated from chemical fumes and vapors for longer life
  • Chemical-resistant glass lid provides visibility of samples and detaches for cleaning
  • Two clamps hold lid securely in place
  • RapidVap models with optional RS-232 link allow all front panel functions to be performed and monitored from personal computer
Includes:
  • 2 m (6.5 ft.) three-wire cord with plug
Specifications:
  • Dimensions w/ closed lid (L x W x H): 46.7 x 52.6 x 34.3 cm (18.4 x 20.7 x 13.5 in.)
  • Weight: 42 kg (92 lb.)
  • 230 V models
  • CE marked
Specifications
Certifications CE
Electrical Rating International/Schuko
Frequency 50/60 Hz
Holds One rotor
Max. Vacuum 999 mbar
Shipping Weight (English) 100 lb.
Shipping Weight (Metric) 45 kg
Type Vacuum dry evaporation system
Voltage 230 V
Volume (English) 0 to 120 oz.
Volume (Metric) 0 to 3600 mL
Temperature Range (Metric) 0°C to 100°C
Chamber Material PTFE-coated Aluminum
Amperage 8 A
Length (English) 18.4 in.
Height (English) 13.5 in.
Height (Metric) 34.3 cm
Length (Metric) 46.7 cm
Speed 1000 rpm
Plug Type Schuko
Weight (English) 100 lb.
Weight (Metric) 42 kg
Width (Metric) 52.6 cm
Width (English) 20.7 in.
Dimensions (L x W x H) 52.6 x 46.7 x 63.2 cm