Learn More
Thermo Scientific™ RapID™ QC Set
Product Code. 12978244
Change view
Click to view available options
Quantity:
5 Loops/Pk.
Product Type:
ANA II QC Set
NF Plus QC Set
NH QC Set
ONE QC Set
SS/u QC Set
STAPH PLUS QC Set
STR QC Set
YEAST PLUS QC Set
Unit Size:
Each
Pack of 5
Product Code. Quantity Product Type unitSize
12978244 5 Loops/Pk. NF Plus QC Set Pack of 5
12915983 5 Loops/Pk. STR QC Set Each
12995973 5 Loops/Pk. ANA II QC Set Each
12905983 5 Loops/Pk. NH QC Set Each
12975973 5 Loops/Pk. STAPH PLUS QC Set Each
12925983 5 Loops/Pk. SS/u QC Set Each
12955983 5 Loops/Pk. YEAST PLUS QC Set Each
12945983 5 Loops/Pk. ONE QC Set Each
8 options
Product Code. 12978244
Brand: Thermo Scientific™ R4653054
Obtain faster recovery, enhanced viability, and direct-streak-to-plate capabilities with QC organisms manufactured using an unmatched proprietary gel preservation process.
Ensure high-performance products, reproducible clinical test results and consistent microbiological evaluation of pharmaceuticals and food with Thermo Scientific™ Culti-Loops™ quality control organisms. Thermo Scientific™ RapID™ QC Sets include a ready-to-use set of QC organisms in Culti-Loops format which are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix.
Trust Thermo Scientific quality control organisms for quality results you can depend on, backed by over 30 years of expertise in the manufacturing and packaging of controlled microorganisms.
Convenience
  • Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration.
Performance
  • Obtain enhanced viability and recovery with proprietary gel-preserved organisms.
Value
  • Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms.
Confidence
  • Reach complete traceability with ATCC™ License Derivative strains.
Efficiency
  • Achieve accurate and convenient documentation with peel-off, transferable labels.
Safety
  • Reduce the risk of contamination and infection with zero organism handling.
  • More than 600 quality control organisms, including convenient sets for commonly used identification systems
  • ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
  • ATCC™ Licensed Derivative Program member
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
*The ATCC Licensed Derivative™ Emblem, the ATCC Licensed Derivative™ word mark, and the ATCC catalog marks are trademarks of ATCC™. Thermo Fisher Scientific is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 13253TM*
Certifications/Compliance ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
Content And Storage 2°C to 8°C
Description RapID™ NF PLUS QC Set/4
Format Culti-Loops
Product Type NF Plus QC Set
Target Organism Class Organism Sets
Test Type Qualitative QC
Packaging Type 5 Loops/Pack
Quantity 5 Loops/Pk.
Species Acinetobacter baumannii, Aeromonas hydrophila, Elizabethkingia meningoseptica, Oligella ureolytica