Learn More
Buerkle™ Rapid Couplings, NW 6.4 mm, Male Connections, Hose Nipples with Hose Nozzles
Product Code. p-7084529 Shop All Bürkle Products
Change view
Click to view available options
Tubing Size:
6.5 mm dia. x 32 mm
6.5 mm dia. x 47 mm
8 mm dia. x 32 mm
9.5 mm dia. x 42 mm
Material:
POM
Polypropylene
Unit Size:
Each
Product Code. Tubing Size Material unitSize
10027062 6.5 mm dia. x 32 mm POM Each
10789664 8 mm dia. x 32 mm POM Each
10086711 9.5 mm dia. x 42 mm Polypropylene Each
10697106 6.5 mm dia. x 47 mm POM Each
10255413 6.5 mm dia. x 47 mm Polypropylene Each
5 options
Product Code. 10027062
Brand: Buerkle™ 87711040
Only null left
Add to basket
Add to basket
Rapid Couplings, NW 6.4 mm, Male Connections, Hose Nipples with Hose Nozzles for a fast and safe connection for liquids, gases, compressed air and vacuum.
  • Rapid couplings are the ideal when connecting and separating hoses, machines, apparatus, containers, etc
  • For a fast and safe connection
  • For liquids, gases, compressed air and vacuum
TRUSTED_SUSTAINABILITY
Specifications
Description Without valve
Diameter (Metric) Inner 6.5 mm
Material POM
Overall Length (Metric) 32 mm
For Use With 6.4 mm Hose
Min. Operating Temperature (Metric) -40°C
Packaging Quantity 1
Tubing Size 6.5 mm dia. x 32 mm
Thread Size NW 6.4 mm
Includes Male connections, hose nipples with hose nozzles
Maximum Pressure 10 bar
Max. Operating Temperature (Metric) 80°C
Product Type Rapid Coupling