Learn More
Buerkle™ Rapid Couplings, NW 6.4 mm, Male Connections, Hose Nipples with Hose Nozzles
Product Code. 10086711 Shop All Bürkle Products
Change view
Click to view available options
Tubing Size:
6.5 mm dia. x 32 mm
6.5 mm dia. x 47 mm
8 mm dia. x 32 mm
9.5 mm dia. x 42 mm
Material:
POM
Polypropylene
Unit Size:
Each
Product Code. Tubing Size Material unitSize
10086711 9.5 mm dia. x 42 mm Polypropylene Each
10027062 6.5 mm dia. x 32 mm POM Each
10789664 8 mm dia. x 32 mm POM Each
10697106 6.5 mm dia. x 47 mm POM Each
10255413 6.5 mm dia. x 47 mm Polypropylene Each
5 options
Product Code. 10086711
Brand: Buerkle™ 87712160
Only null left
Add to basket
Add to basket
Rapid Couplings, NW 6.4 mm, Male Connections, Hose Nipples with Hose Nozzles for a fast and safe connection for liquids, gases, compressed air and vacuum.
  • Rapid couplings are the ideal when connecting and separating hoses, machines, apparatus, containers, etc
  • For a fast and safe connection
  • For liquids, gases, compressed air and vacuum
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Overall Length (Metric) 42 mm
For Use With 9.5 mm Hose
Packaging Quantity 25/Scale
Tubing Size 9.5 mm dia. x 42 mm
Width (Metric) 6.4 mm
Includes Valve
Maximum Pressure 8 bar
Product Type Rapid Coupling