Learn More
STEMCELL Technologies Rapamycin
Description
- Rapamycin is a macrolide antibiotic and immunosuppressive compound that inhibits mammalian target of rapamycin (mTOR) signaling
- It acts through formation of a complex with cytosolic FK-binding protein 12 (FKBP-12), which directly binds to mTOR complex 1 (mTORC1)
- Its immunosuppressive effects are mediated through inhibition of IL-2 signaling that is critical for T-cell proliferation and activation (Gibbons et al.; Kay et al.) Rapamycin shows antifungal activity against Candida albicans and other fungi (Vézina et al.)
- Inhibits growth of MDA-MB-468 human breast cancer cells in vitro, and inhibits tumor growth in a mouse xenograft model in vivo (Akcakanat et al.)
- Induces autophagy in malignant glioma cells (Takeuchi et al.)
Specifications
Specifications
| CAS | 53123-88-9 |
| Quantity | 1 mg |
| Percent Purity | ≥95% |