missing translation for 'onlineSavingsMsg'
Learn More

RAP2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7207338 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16895844 20 μL 20µL
16885844 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16895844

Brand: Proteintech 162661AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This intronless gene belongs to a family of RAS-related genes. The proteins encoded by these genes share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between the RAP and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. Evidence suggests that this protein may be polyisoprenylated and palmitoylated.

Specifications

Antigen RAP2B
Applications Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAP2B
Gene Accession No. P61225, P61226, P61227
Gene Alias RAP2B, Ras related protein Rap 2b
Gene Symbols Rap2b
Host Species Rabbit
Immunogen RAP2B Fusion Protein Ag9331
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170923, 5912, 74012
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.