missing translation for 'onlineSavingsMsg'
Learn More

RAP1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10840-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847493

  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RAP1B and RAP1A belong to a superfamily of RAS like small GTP-binding proteins involved in cell signaling.
Specifications

Specifications

RAP1B
Polyclonal
Unconjugated
RAP1B
GTP binding protein smg p21B, K REV, RAL1B, RAP1B, Ras related protein Rap 1b
Rabbit
Antigen Affinity Chromatography
RUO
171337, 215449, 5908
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P61224, Q62636, Q99JI6
Rap1b
RAP1B Fusion Protein Ag1282
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers