missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RAP1A Recombinant Protein

Brand:  Abnova™ H00005906-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190042

  • 4115.00 NOK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH14086.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
45.98
8
Glutathione Sepharose 4 Fast Flow
10 ug
MREYKLVVLGSGGVGKSALTVQFVQGIFVEKYDPTIEDSYRKQVEVDCQQCMLEILDTAGTEQFTAMRDLYMKNGQGFALVYSITAQSTFNDLQDLREQILRVKDTEDVPMILVGNKCDLEDERVVGKEQGQNLARQWCNCAFLESSAKSKINVNEIFYDLVRQINRKTPVEKKKPKKKSCLLL
KREV-1/KREV1/RAP1/SMGP21
RAP1A
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
5906
RAP1A (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RAP1A
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers