Learn More
RAP1A Mouse anti-Human, Mouse, Rat, Clone: 1G12D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68125-1-IG-150UL
This item is not returnable.
View return policy
Description
RAP1A Monoclonal antibody specifically detects RAP1A in Human, Mouse, Rat samples. It is validated for IHC, WB, ELISASpecifications
| RAP1A | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Mouse | |
| 150 μL | |
| 5906 | |
| Store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, ELISA | |
| 1G12D8 | |
| Unconjugated | |
| BC001559 | |
| Protein A purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |