Learn More
Brannan™ Range Finder Hydrometers
Description
Brannan Range Finder Hydrometers, Baume Range: 1.50 to 2, Length: 250mm, Certifications/Compliance: UKAS, Quantity: 1Each, Subdivisions: 0.005g/mL
Specifications
Specifications
| Quantity | 1Each |
| Type | Range Finder Hydrometer |
| Increments | 0.005 g/mL |
| Certifications/Compliance | UKAS |
| Length (Metric) | 250 mm |
| Baume Range | 1 to 1.50 |
| For Use With (Application) | Specific Gravity Measurement |
| Specific Gravity Range | 1 to 1.5 |