missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17656-1-AP-20UL
This item is not returnable.
View return policy
Description
RANBP6 (Ran-binding protein 6) belongs to the importin beta family and may function in nuclear protein import as a nuclear transport receptor.Specifications
| RANBP6 | |
| Polyclonal | |
| Unconjugated | |
| RANBP6 | |
| RAN binding protein 6, RANBP6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 240614, 26953 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60518, Q8BIV3 | |
| RANBP6 | |
| RANBP6 Fusion Protein Ag11906 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction