Learn More
Thermo Scientific™ MarqMetrix Replacement Process BallProbe
Product Code. 17739417 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Replacement Process BallProbe
Unit Size:
Each
Product Code. Type unitSize
17739417 Replacement Process BallProbe Each
1 options
Product Code. 17739417
Brand: Thermo Scientific™ 8100284101
This item requires a price update prior to being ordered.
Request a Price for this item here.
Replacement BallProbe for use with the MarqMetrix All-In-One process raman analyzer.
  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MarqMetrix All-In-One Process Raman Analyzer
Type Replacement Process BallProbe