Learn More
Thermo Scientific™ MarqMetrix Replacement Fiberhead BallProbe
Shop All Thermo Scientific Products
Click to view available options
Type:
Replacement Fiberhead BallProbe
Unit Size:
Each
Description
- Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
Specifications
Specifications
| For Use With (Equipment) | MarqMetrix All-In-One Process Raman Analyzer |
| Type | Replacement Fiberhead BallProbe |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering