Learn More
Thermo Scientific™ MarqMetrix Replacement Bioreactor BallProbe
Product Code. 17749417 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
MarqMetrix All-In-One Process Raman Analyzer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17749417 MarqMetrix All-In-One Process Raman Analyzer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17749417 Supplier Thermo Scientific™ Supplier No. 8100285101
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Replacement BallProbe for use with the MarqMetrix All-In-One process raman analyzer.
  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Bioreactor Ball Probe
For Use With (Equipment) MarqMetrix All-In-One Process Raman Analyzer