missing translation for 'onlineSavingsMsg'
Learn More

RALYL Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17179-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847024

  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RALYL belongs to the RRM HNRPC family, RALY subfamily. It contains 1 RRM (RNA recognition motif) domain. The functions of RALYL remain unknown.
Specifications

Specifications

RALYL
Polyclonal
Unconjugated
RALYL
hnRNP core protein C like 3, HNRPCL3, hRALYL, RALY RNA binding protein like, RALYL, RNA binding Raly like protein
Rabbit
Antigen Affinity Chromatography
RUO
138046, 76897
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86SE5, Q8BTF8
Ralyl
RALYL Fusion Protein Ag10898
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers