Learn More
Raloxifene Hydrochloride, Mikromol™
Description
Raloxifene Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Raloxifene hydrochloride |
| CAS | 82640-04-8 |
| Analyte or Component Names | Raloxifene hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H27 N O4 S . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Raloxifene Hydrochloride |
| InChI Formula | InChI=1S/C28H27NO4S.ClH/c30-21-8-4-20(5-9-21)28-26(24-13-10-22(31)18-25(24)34-28)27(32)19-6-11-23(12-7-19)33-17-16-29-14-2-1-3-15-29;/h4-13,18,30-31H,1-3,14-17H2;1H |
| SMILES | Cl.Oc1ccc(cc1)c2sc3cc(O)ccc3c2C(=O)c4ccc(OCCN5CCCCC5)cc4 |
| IUPAC Name | [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-1-ylethoxy)phenyl]methanone;hydrochloride |
| Show More |