missing translation for 'onlineSavingsMsg'
Learn More

RALGAPB Rabbit anti-Human, Rat, Mouse, Polyclonal, Proteintech

Product Code. 17659626 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17659626 150 μL 150µL
17669626 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 17659626

Brand: Proteintech 283301AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RALGAPB Polyclonal antibody apecifically detetects RALGAPB antigen in Human, Rat, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.

Specifications

Antigen RALGAPB
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 550 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC047482
Gene Symbols KIAA1219
Host Species Rabbit
Immunogen RALGAPB fusion protein Ag28259
Purification Method Antigen affinity purification
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 57148
Target Species Human, Rat, Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.