missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALGAPB Rabbit anti-Human, Rat, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28330-1-AP-20UL
This item is not returnable.
View return policy
Description
RALGAPB Polyclonal antibody apecifically detetects RALGAPB antigen in Human, Rat, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| RALGAPB | |
| Polyclonal | |
| Unconjugated | |
| BC047482 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human, Rat, Mouse | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 550 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| KIAA1219 | |
| RALGAPB fusion protein Ag28259 | |
| 20 μL | |
| 57148 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction